Learn More
MSC Brady Laboratory Labels, Cryogenic Polypropylene, Chemical Resistant, 1 cartridge
Catalog No. 5000677438
Encompass
These Brady B-7425 polypropylene labels are chemical and temperature resistant, designed for harsh lab processes. They enable easy text reading and barcode scanning for laboratory identification on vials, centrifuge tubes, and test tubes, maintaining adhesion under challenging conditions.
  • Withstand extreme conditions for laboratory identification
  • Resist chemicals, liquid nitrogen, and other harsh environments
  • Temperature resistance from -112°F to 158°F (-80°C to 70°C)
  • Permanent acrylic adhesive for strong bonding to smooth surfaces
  • Autoclavable, cryogenic storage, low temperature, and permanent adhesive properties
  • Resistant to chemicals, cold, high/low temperatures, smear, and solvents
  • RoHS compliant
  • Designed for indoor use
  • Compatible with BMP21, IDPAL, LABPAL, and M210/M211 series printers
  • Print technology: thermal transfer
Catalog No. 50-006-77438 Supplier MSC Supplier No. 98904881
These Brady B-7425 polypropylene labels are chemical and temperature resistant, designed for harsh lab processes. They enable easy text reading and barcode scanning for laboratory identification on vials, centrifuge tubes, and test tubes, maintaining adhesion under challenging conditions.
  • Withstand extreme conditions for laboratory identification
  • Resist chemicals, liquid nitrogen, and other harsh environments
  • Temperature resistance from -112°F to 158°F (-80°C to 70°C)
  • Permanent acrylic adhesive for strong bonding to smooth surfaces
  • Autoclavable, cryogenic storage, low temperature, and permanent adhesive properties
  • Resistant to chemicals, cold, high/low temperatures, smear, and solvents
  • RoHS compliant
  • Designed for indoor use
  • Compatible with BMP21, IDPAL, LABPAL, and M210/M211 series printers
  • Print technology: thermal transfer