Learn More
Brady™ LABXPERT™ and IDXPERT™ Vial Top Labels
Catalog No. 22500546 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, White
Catalog No. Color
22-500-546 Black, White
1 options
Catalog No. 22-500-546 Supplier Brady™ Supplier No. X83499
Round nylon cloth labels for use with Brady LABXPERT and IDXPERT printers
  • Withstands extremely low temperatures
  • Excellent water and oil resistance
  • Good moisture resistance
  • High adhesion with permanent acrylic adhesive
  • Various colors available
UL recognized with its respective ribbon
General and industrial identification, inside-of-panel component marking, laboratory identification, panel identification, vial, vial top
Compliance
Compliant with RoHS Directive
Specifications
Certifications/Compliance UL
Adhesive Type Acrylic, Permanent
Color Black, White
Format Cartridge (Label)
Material Nylon Cloth
Product Type Laboratory Label
For Use With (Application) General and industrial identification, inside of panel component marking, laboratory identification, panel identification, vial, vial top
Quantity 300/Pk.
Width (English) 0.5 in.
Height (English) 0.5 in.
Height (Metric) 1.2 cm