Learn More
Brady™ Large Spill Truck, Oil Only
Catalog No. 17111274 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
115 gal./Bale
Catalog No. Capacity (English)
17-111-274 115 gal./Bale
1 options
Catalog No. 17-111-274 Supplier Brady™ Supplier No. SKOLT
  • $2,890.00
    $2,250.65 / Each of 1
    Save $639.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Includes
200 pads (15 x 19 in.), 24 SOCs (3 in. x 4 ft.), 12 SOCs (3 in. x 12 ft.), 8 pillows (17 x 19 in.), 15 disposal bags, 1 pair goggles, 3 pair nitrile gloves, Emergency Response Guidebook
For use with oil based materials
  • Made of polyethylene
  • Chemical and water resistant
  • Wheel mounted for mobility
  • Absorbs up to 115 gal./bale
  • Not suitable for outdoor use
Specifications
Capacity (English) 115 gal./Bale
Material Polyethylene
Type Spill Truck Kit, Oil-only
Includes (200) 15 x 19 in. Pads, 3 in. x 4 ft. SOCs (24), 3 in. x 12 ft. SOCs (12), (8) 17 x 19 in. pillows, (3) pair nitrile gloves, (15) disposal bags, goggles, handbook
Size Large
Weight (English) 184.5 lb.
Weight (Metric) 406.82 kg