Learn More
Brady™ Large Truck Spill Kit Refill, Universal
Catalog No. 17111253 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
134 gal.
Catalog No. Capacity (English)
17-111-253 134 gal.
1 options
Catalog No. 17-111-253 Supplier Brady™ Supplier No. SKALTR
  • $1,106.00
    $861.65 / Each of 1
    Save $244.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Large Truck Spill Kit Refill is intended for use with the Large Spill Truck, Universal. (Refill kit only—container sold separately.)
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
Specifications
Color Gray
Capacity (English) 134 gal.
Capacity (Metric) 507.2 L
Type Universal Spill Kit Refill
Includes 200 pads (15 x 19 in.), 24 SOCs (3 in. x 4 ft.), 12 SOCs (3 in. x 12 ft.), 8 pillows (17 x 19 in.), 15 disposal bags, 1 pair goggles, 3 pair nitrile gloves, emergency response guidebook