Learn More
MSC Brady Lockout Hasp, Scissor-Action, Single Jaw, 6 Padlocks, 1 ea
Catalog No. 5000684984
Encompass
This lockout hasp features a scissor-action design with a single jaw, capable of securing up to 6 padlocks. Constructed from steel, it is designed for effective energy isolation during maintenance or repair operations.
  • Scissor-action design
  • Single jaw configuration
  • Accommodates up to 6 padlocks
  • Made from durable steel
  • Case-hardened with rust-proof plating
  • Bright red vinyl insulation for high visibility
Catalog No. 50-006-84984 Supplier MSC Supplier No. 28548733
This lockout hasp features a scissor-action design with a single jaw, capable of securing up to 6 padlocks. Constructed from steel, it is designed for effective energy isolation during maintenance or repair operations.
  • Scissor-action design
  • Single jaw configuration
  • Accommodates up to 6 padlocks
  • Made from durable steel
  • Case-hardened with rust-proof plating
  • Bright red vinyl insulation for high visibility