Learn More
MSC Brady Lockout Hasp, Scissor-Action, Single Jaw, 6 Padlocks, 1 ea
Catalog No. 5000793221
Encompass
This lockout hasp features a scissor-action design and a single jaw, allowing for the securing of machinery and energy sources. It can accommodate up to six padlocks, ensuring comprehensive lockout/tagout procedures are followed.
  • Scissor-action hasp type
  • Single jaw design
  • Accommodates up to 6 padlocks
  • Constructed from steel
  • Red color for high visibility
  • Case-hardened steel with rust-proof plating
  • Bright red vinyl insulation
Catalog No. 50-007-93221 Supplier MSC Supplier No. 68450873
This lockout hasp features a scissor-action design and a single jaw, allowing for the securing of machinery and energy sources. It can accommodate up to six padlocks, ensuring comprehensive lockout/tagout procedures are followed.
  • Scissor-action hasp type
  • Single jaw design
  • Accommodates up to 6 padlocks
  • Constructed from steel
  • Red color for high visibility
  • Case-hardened steel with rust-proof plating
  • Bright red vinyl insulation