Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Marking Flag

Catalog No. 19103881 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-881 Black on Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-881 Supplier Brady™ Supplier No. 98168
  • $60.50
    $44.65 / Pack of 100

    Save $15.85

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for identifying possible hazards buried underground

  • Custom legends available
  • Attached to 76.2cm (30 in.), 15.5 ga steel rod

Specifications

Color Black on Red
Width (Metric) 12.7 cm
Width (English) 5 in.
Description Legend: Buried Electric Line
Height (English) 4 in.
Height (Metric) 10.1 cm
Packaging Quantity 100/Pk.
Material Plastic flag, Steel rod
Type Marking Flag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.