Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads
GSA_VA

Catalog No. 19150260 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
22 gal.
28 gal.
Weight Capacity:
Heavyweight
Lightweight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Weight Capacity
19-150-260 22 gal. Heavyweight
19-103-101 28 gal. Lightweight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-150-260 Supplier Brady™ Supplier No. CH100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color-coded absorbents for safe clean-up of hazardous fluids. Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads are surfactant treated polypropylene pads used on wide range of chemicals.

  • Benefits of sign and an absorbent in one
  • Highly absorbent for all-purpose industrial applications
  • Ideal for areas where the risk of slips, trips and falls are greatest
  • Chemically inert, do not react with aggressive fluids
  • Perforations provide cost control and reduce waste
  • Heavy weight - specifically designed for continuous leaks and larger spills
  • Light weight - specifically designed for smaller leaks, drips and spills

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
Color Black/Red on Yellow
Description Does Not Contain Recycled Content
Capacity (English) 22 gal.
Capacity (Metric) 83.3 L
Material Polypropylene
Type Perforated Absorbent Pad
Packaging Quantity 100/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 19 x 15 in. (48.2 x 38 cm)
Length (English) 19 in.
Length (Metric) 48.2 cm
Width (English) 15 in.
Width (Metric) 38 cm
Weight (English) 15 lb.
Weight (Metric) 6.8 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.