Learn More
Brady™ Mini Cable Lockout with Metal Cable
Catalog No. 19000255 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Mini Cable Lockout
Catalog No. Product Type
19-000-255 Mini Cable Lockout
1 options
Catalog No. 19-000-255 Supplier Brady™ Supplier No. 50940
  • $91.00
    $51.65 / Each of 1
    Save $39.35
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Mini Cable Lockout with Metal Cable is used for locking out gate valves, handles and other oversized devices.
  • Mini Cable Lockouts are extremely compact and portable, cable stored internally
  • Push button self-winding mechanism retracts automatically to tightly cinch the cable, leaving no slack
  • Device comes complete with a sheathed metal lockout cable attachment
  • Superior chemical, corrosion and temperature-resistant properties
Specifications
Product Type Mini Cable Lockout
Certifications/Compliance OSHA 1910.147