Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Mini Gate Valve Lockout

Catalog No. 19037914 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-037-914 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-037-914 Supplier Brady™ Supplier No. 65590
  • $53.75
    $32.65 / Each of 1

    Save $21.10

    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mini Gate Valve Lockout devices deny access and cover the valve handle. The unique knockout center fits OS&Y or rising stem valves.

  • Device is made of extremely rugged injection-molded polypropylene for superior impact and chemical resistance
  • Supplied with English, Spanish and French danger labels
  • Cover the valve handle with the gate valve lockout device and apply a personal padlock and lockout tag to ensure employee safety
  • Accepts padlock shackles up to 3/8 in. in diameter

Specifications

Product Type Mini Gate Valve Lockout
Color Yellow
Material Polypropylene
Certifications/Compliance OSHA 1910.147
Description Fits Valve Wheel Diameter 1 - 2.5 in. (3 to 6 cm)
Diameter (English) 2.78 in.
Diameter (Metric) 7.06 cm
Temperature Range (English) −20° to +80°F
Temperature Range (Metric) 0° to 175°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.