Learn More
Brady™ MRO™ Plus Medium Rolls
Catalog No. 17111194 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
20 gal./Bale
Catalog No. Capacity (English)
17-111-194 20 gal./Bale
1 options
Catalog No. 17-111-194 Supplier Brady™ Supplier No. MRO315P
  • $183.00
    $127.65 / Each of 1
    Save $55.35
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)
  • Three-ply meltblown polypropylene construction
  • Perforated - reduces waste
  • Gray color masks drips and spills
  • Available in various sizes
Specifications
Capacity (English) 20 gal./Bale
Material Polypropylene
Type MRO Plus Medium Roll
For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up, drums - liquid storage, outfalls and discharge ponds
Packaging Quantity 1 Ea.
Dimensions (L x W) 45.72 m x 76.2 cm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov