Learn More
Brady™ MRO Plus™ Medium Rolls
Catalog No. 17111202 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
38 gal./Bale
Catalog No. Capacity (English)
17-111-202 38 gal./Bale
1 options
Catalog No. 17-111-202 Supplier Brady™ Supplier No. MRO350DP /DEL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)
  • Three-ply meltblown polypropylene construction
  • Perforated - reduces waste
  • Gray color masks drips and spills
  • Available in various sizes
Specifications
Capacity (English) 38 gal./Bale
Type MRO Plus Medium Roll
For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up, drums - liquid storage, outfalls and discharge ponds
Packaging Quantity 1 Ea.
Weight Capacity Heavyweight
Dimensions (L x W) 38.1 x 45.72 cm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov