Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Oil Only Drum Top Cover

Catalog No. 19000158 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Drums
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-000-158 Drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-000-158 Supplier Brady™ Supplier No. DTO25
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oil Only Drum Top Cover keeps drums, floor, and surrounding storage areas clean.

  • Drum top covers help to maintain clean work surfaces by preventing pump spills and overspray from reaching the floor
  • White absorbents pick up petroleum-based fluids, paints and non-water soluble chemicals while repelling water - great for indoor and outdoor use
  • Oil Only absorbents are unaffected by temperature and impervious to rot and mildew
  • Floats indefinitely even when saturated with oil

Specifications

Product Type Oil Only Drum Top Cover
Color White
For Use With (Application) Overspray Leaks and Drips, Workstations, Drums - Liquid Storage, Outfalls and Discharge Ponds
For Use With (Equipment) Drums
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.