Learn More
Brady™ Oil Plus™ Heavy Pad, Dispense-n-Dispose
Catalog No. 17111133 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
39 gal./Bale
Catalog No. Capacity (English)
17-111-133 39 gal./Bale
1 options
Catalog No. 17-111-133 Supplier Brady™ Supplier No. OP150DND
  • $230.00
    $167.65 / Case of 150
    Save $62.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Oil Plus™ Dispense-n-Dispose heavy pads absorb petroleum-based oils, solvents and other non-water soluble liquids.
  • White absorbents pick up petroleum-based fluids, paints and non-water soluble chemicals while repelling water - great for indoor and outdoor use
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • 3-ply meltblown polypropylene construction provides durability; pads float indefinitely even when saturated with oil
  • Perforations provide cost control and reduce waste by allowing you to use only what you need
Specifications
Color White
Capacity (English) 39 gal./Bale
Material Polypropylene
Type Oil Plus Heavy Pad, Dispense-n-Dispose
For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up, drums - liquid storage, outfalls and discharge ponds, heavy manufacturing, indoor/outdoor petroleum clean-up
Packaging Quantity 150/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 38.1 x 48.2 cm (15 x 19 in.)
Length (Metric) 48.26 cm
Width (Metric) 38.1 cm