Learn More
Brady™ Oil Spill Control Absorbent Storage Cart Fill Kit
Catalog No. 17111004 Shop All Brady Corporation Products
Change view
Click to view available options
Oil Spill Control Absorbent Storage Cart Fill Kit
Catalog No.
17-111-004 Oil Spill Control Absorbent Storage Cart Fill Kit
1 options
Catalog No. 17-111-004 Supplier Brady™ Supplier No. SCCARTOF
  • $556.00
    $412.65 / Each of 1
    Save $143.35
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Absorbent Storage Cart is designed to provide storage and enable quick access to all of the essential absorbents for addressing oil spills.
  • Spill Control Center Refill: Oil Only, 40 gal
Specifications
Color White
Type Oil Spill Control Absorbent Storage Cart Refill Kit
Includes 15 x 19 in. pads (100), 3 in. x 4 ft. socks (8), 8 pillows, 5 disposal bags, 1 pair Nitrile gloves, goggles