Description
- This compact kit comes with a belt pouch for easy access and transportation
- The Personal Padlock Pouch Kit includes two keyed-alike padlocks, two hasps, two tags with cable ties, and a belt pouch
- Assigning keyed-alike padlocks to each employee simplifies key management and ensures that employees can quickly and easily find the right key
- Remember, to comply with safety requirements, a set of keyed-alike padlocks should never be split among multiple employees
Specifications
Specifications
| Product Type | Padlock Kit with Keyed-Alike Safety Padlocks |
| Color | Red |
| Certifications/Compliance | OSHA 1910.147 |
| Includes | Two Keyed-alike Safety Padlocks, Two 1 in. Group Lockout Hasps, Two Heavy Duty Lockout Tags with Nylon Cable Ties, One Lockout Belt Pouch |
| Material (Shackle) | Steel |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.