Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biohazard Safety Poster

Catalog No. 19103293 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Laminated paper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-103-293 Laminated paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-293 Supplier Brady™ Supplier No. PS125E

Contains universal precautions for those occupationally exposed to blood or other potentially infectious materials

  • Laminated on both sides for extra durability

Specifications

Material Laminated paper
Quantity 1 Ea.
Height (English) 18 in.
Height (Metric) 45.7 cm
Width (English) 24 in.
Width (Metric) 61 cm
Displayed Text Bloodborne pathogens
Product Type Biohazard safety poster
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.