Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Haz-Mat Warning Labels Poster

Catalog No. 19103295 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Paper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-103-295 Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-295 Supplier Brady™ Supplier No. PS135E
  • $86.00
    $80.65 / Each of 1

    Save $5.35

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haz-Mat Warning Labels Poster is a label guide for hazardous chemicals used in the area.

  • Easy-to-read, large format poster gets attention
  • Lamination protects graphics and improves durability
  • Use with optional clear acrylic poster holder (SH1824, sold separately) to store different posters and rotate them according to your safety program
  • Helps reduce accidents and increase facility safety through employee education

Specifications

Material Paper
Quantity 1 Ea.
Mounting Type Wall Mount
Height (English) 18 in.
Height (Metric) 45.7 cm
Width (English) 24 in.
Width (Metric) 61 cm
Shape Rectangular
Languages English
Dimensions (L x W) 18 x 24 in.
Product Type Hazardous Materials Identification Poster
Color Yellow on White
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.