Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Label Hazard Id Biohazard: Biohazard Infectious Agent

Catalog No. 19140978 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-140-978 Black, Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-140-978 Supplier Brady™ Supplier No. LH625E
  • $49.15
    $38.65 / Pack of 25

    Save $10.50

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biohazard warning label reads Biohazard: Infectious Agent with Pictogram. Packs of 25 black-on-orange labels.

  • An essential step towards safety
  • Clear and instantly-readable identification of biohazard areas and waste products

Specifications

Adhesive Type Permanent Acrylic
Color Black, Orange
Finish Gloss
Format Label
Material Vinyl
Printable Sides Single-side Printable
Product Type Biohazard Warning Tape
Sterility Non-sterile
Display Text Biohazard Infectious Agent
Dimensions (L x W) 4 x 2.875 in.
Width (English) 2.875 in.
Width (Metric) 73 mm
Height (English) 4 in.
Height (Metric) 101 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.