Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Reinforced Barricade Tapes

Catalog No. 19104311 Shop All Brady Corporation Products
Change view
Click to view available options
Displayed Text:
Caution
Caution Do Not Enter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
19-104-311 Caution Do Not Enter
19-104-309 Caution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-104-311 Supplier Brady™ Supplier No. 91102
  • $207.00
    $129.65 / Each of 1

    Save $77.35

    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides fast, on-the-spot warning

  • Provides long-lasting hazard protection even with long distances between tie points
  • Tough enough to be used again and again without stretching or sagging in sunlight or wind
  • Standard package: One roll
  • Offers 113.4kg (250 lb.) tensile strength
  • Material thickness is 4mil

Specifications

Color Black, Yellow
Width (English) 3 in.
Width (Metric) 7.62 cm
Length (English) 500 ft.
Length (Metric) 152.4 m
Displayed Text Caution Do Not Enter
Type Reinforced Barricade Tape
Dimensions (L x W) 152.4 m x 7.62 cm (500 ft. x 3 in.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.