Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Right-To-Know Label Blanks

Catalog No. 19807127 Shop All Brady Corporation Products
Change view
Click to view available options
Width (English):
2 in.
5 in.
7 in.
Height (English):
2 in.
3.5 in.
5 in.
Product Type:
Blank RTK Label (Color Bar)
Blank RTK Label (Diamond Hazard)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Height (English) Product Type
19-807-127 5 in. 3.5 in. Blank RTK Label (Diamond Hazard)
19-807-126 5 in. 3.5 in. Blank RTK Label (Color Bar)
19-807-128 2 in. 2 in. Blank RTK Label (Diamond Hazard)
19-807-125 7 in. 5 in. Blank RTK Label (Color Bar)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-807-127 Supplier Brady™ Supplier No. 59254

Blank RTK labels featuring a permanent, pressure-sensitive adhesive backing

  • Compatible with laser printers only
  • Available with color bar design or diamonds with hazard classification

Specifications

Adhesive Type Permanent, Pressure-sensitive
Format Label
Material Vinyl
Product Type Blank RTK Label (Diamond Hazard)
For Use With (Application) Used for safety, maintenance and general markings within a facility
For Use With (Equipment) Laser printer
Life Span 3 to 5 Years
Quantity 25/Pk.
Temperature Range (English) -40 to 180°F
Temperature Range (Metric) -40°C to +82°C
Tensile Strength 14.4 lb./in.
Thickness (English) 0.0045 in.
Width (English) 5 in.
Height (English) 3.5 in.
Height (Metric) 8.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.