Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SDS Elements Signs

Catalog No. 19000477 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Fiberglass
Polyester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-000-477 Fiberglass
19-000-480 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-000-477 Supplier Brady™ Supplier No. 133169

To identify equipment, rooms and materials, which contain or are contaminated with hazardous agents

  • Available in fiberglass and self-sticking polyester material
  • Size: 25.4W x 35.5cm H(10 x 14 in.)

Specifications

Material Fiberglass
Quantity 1 Ea.
Description Fiberglass (B-140)
Height (English) 14 in.
Height (Metric) 35.5 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Dimensions (L x W) 25.4 x 35.5cm
Displayed Text SDS Format
Product Type SDS Elements Sign
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.