Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Self-Laminating Danger Tags with Different Fronts and Backs

Catalog No. 19040801 Shop All Brady Corporation Products
Change view
Click to view available options
Message:
Header: DANGER, Front: DO NOT OPERATE / SIGNED BY / DATE / PHONE, Back: DO NOT REMOVE THIS TAG / Remarks
Header: DANGER, Front: OUT OF SERVICE / SIGNED BY / DATE / PHONE, Back: DO NOT REMOVE THIS TAG / Remarks
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Message
19-040-801 Header: DANGER, Front: DO NOT OPERATE / SIGNED BY / DATE / PHONE, Back: DO NOT REMOVE THIS TAG / Remarks
19-040-802 Header: DANGER, Front: OUT OF SERVICE / SIGNED BY / DATE / PHONE, Back: DO NOT REMOVE THIS TAG / Remarks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-040-801 Supplier Brady™ Supplier No. 86458
Only null left
Add to Cart
Add to Cart

Includes

Nylon fasteners

Personalize with special instructions, hazards, or warnings

  • Protect and seal personalized messages with clear, 1 mil polyester adhesive overlay
  • 0.38 in. (0.95cm) grommeted hole

Order Info

Dimensions (L x W): 5.75 x 3 in. (14.6 x 7.6cm).

Specifications

Certifications/Compliance ASTM D1593, ASTM D638
Color Black, Red, White
Material Polyester
Message Header: DANGER, Front: DO NOT OPERATE / SIGNED BY / DATE / PHONE, Back: DO NOT REMOVE THIS TAG / Remarks
For Use With (Application) For marking on-the-spot warning of dangerous conditions
Durability 3 to 5 Years
Height (English) 5.75 in.
Height (Metric) 14.6cm
Includes 10 nylon ties
Thickness 0.36 mm
Temperature Range (Metric) -40°C to 120°C
Temperature Range (English) -40°F to 248°F
Width (English) 3 in.
Width (Metric) 7.62 cm
Diameter (English) Hole 0.37 in.
Diameter (Metric) Hole 0.93 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.