Learn More
Brady™ Shadow Vinyl Rolls
Catalog No. p-6570005 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-835 Red
19-103-836 Blue
19-103-837 Yellow
19-103-838 Green
19-103-839 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-103-835 Supplier Brady™ Supplier No. 121117
Only null left
Add to Cart
Add to Cart
Allows to quickly and easily create tool shadows to exact size and shape
  • Place tool on vinyl, trace outline with pen and cut out with scissors
Specifications
Dimensions (L x W) 12 ft. x 12 in.
Color Red
Adhesive Type Permanent Pressure Sensitive Acrylic
Material Vinyl
Type Shadow Roll