Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Sign Base

Catalog No. 19054914 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-054-914 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-054-914 Supplier Brady™ Supplier No. 86339
  • $596.00
    $459.65 / Each of 1

    Save $136.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sign Base can be mounted on sidewalks or paved areas without costly digging and damage.

  • Cast aluminum base is 15 in. in diameter and includes post mount stub and hardware
  • Mounts securely to surface using three expansion bolts (not included)
  • Accepts U-channel sign posts (sold separately)
  • Ideal for temporary use

Specifications

Includes Post mount stub and hardware
Material Aluminum
Quantity 1 Ea.
Description Cast Aluminum
Diameter (English) 15 in.
For Use With (Application) Sign base can be mounted on sidewalks or paved areas without costly digging and damage
Product Type Sign Base
Color Silver

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.