Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Signs: ELECTRICAL (W/PICTO)

Catalog No. 19109747 Shop All Brady Corporation Products
Catalog No. 19-109-747 Supplier Brady™ Supplier No. 88497

ELECTRICAL (with PICTOGRAM) signs identify electrical hazard and high voltage areas throughout a facility.

  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)

Specifications

Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Material Polyester
Adhesive Type Permanent Acrylic
Height (English) 10 in.
Thickness 0.01 in.
Width (Metric) 25.4 cm
Dimensions (L x W x H) 10 in. H x 10 in. W x 0.006 in. D
Color Black on Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.