Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Signs: Emergency Stairs

Catalog No. 19150824 Shop All Brady Corporation Products
Catalog No. 19-150-824 Supplier Brady™ Supplier No. 104496

missing translation for 'validMainframeMsg'

EMERGENCY STAIRS ToughStripe™ Floor Signs feature a highly visible, glossy appearance for communicating important safety messages and anti-skid sections to help prevent slips, trips and falls.

  • ToughStripe™ Floor Signs communicate important safety and warning messages to your employees in areas with foot traffic
  • Durable ToughStripe™ Anti-Skid Polyester (B-534) includes a rigid, clear overlaminate and is selectively covered by anti-skid tread
  • Highly visible, glossy material resists harsh chemicals and is easy to apply and remove
  • Floor signs alert workers of potential hazards, direct traffic, and enforce protective equipment requirements.

Specifications

Certifications/Compliance ANSI A1264.2-2006, OSHA 1910.22, ASTM ANSI F1679
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Rubber-based
Description ToughStripe - Pre-printed Floor Signs
Diameter (English) 17 in.
Diameter (Metric) 431.80 mm
For Use With (Application) To alert workers of potential dangers
Max. Temperature (English) 130°F
Min. Temperature (English) 0°F
Thickness 3 mil
Temperature Range (Metric) -18°C to 54°C
Shape Round
Temperature Range (English) 0°F to 130 °F
Languages English
Displayed Text Emergency Stairs
Product Type Floor Sign
Color Black on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.