Description
- Conform heat-shrinkable markers tightly to wire
- Deflector shield evenly distributes forced air heat
- Requires minimum temperature 260°C (500°F) around sleeves of up to 1.9cm (0.75 in.) diameter for uniform shrinkage
- Sturdy base with rubber pad allows hands-off operation
Specifications
Specifications
| Min. Temperature (English) | 500°F |
| Min. Temperature (Metric) | 260°C |
| Product Type | Heat Gun |