Learn More
Brady™ Slow Moving Vehicle Sign
Catalog No. 19104348 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
19-104-348 Steel
1 options
Catalog No. 19-104-348 Supplier Brady™ Supplier No. 57894
  • $32.90
    $22.65 / Each of 1
    Save $10.25
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For vehicles traveling 25mph or less
  • Improves safety
  • Two holes located on back of the sign: First is 10.1cm (4 in.) from top, Second 10.1cm (4 in.) from bottom
  • Holes are not visible on front of the sign
  • Hardware not included
  • Size: W x H: 40.6 x 35.5cm (16 x 14 in.)
Specifications
Material Steel
Type Slow moving vehicle sign