Learn More
MSC Brady SPC Absorbent Roll, Heavy, 30 in W, 150 ft
Catalog No. 5000792712
Encompass
BrightSorb High Visibility absorbents feature bright safety yellow coloring for easy separation of hazardous waste. They are ideal for areas with a high risk of slips, trips, and falls. This surfactant-treated absorbent picks up a wide range of aggressive chemicals while remaining chemically inert, specifically designed for continuous leaks and larger spills.
  • Reduces material usage with heavy weight design for continuous leaks and larger spills
  • Integrates safety signage and absorbency with bright high-visibility color
  • Allows for cost control and waste reduction through perforations
  • Warns of slips, trips, and falls with pre-printed message; bright color alerts to leaks even in low light
Catalog No. 50-007-92712 Supplier MSC Supplier No. 84371749
BrightSorb High Visibility absorbents feature bright safety yellow coloring for easy separation of hazardous waste. They are ideal for areas with a high risk of slips, trips, and falls. This surfactant-treated absorbent picks up a wide range of aggressive chemicals while remaining chemically inert, specifically designed for continuous leaks and larger spills.
  • Reduces material usage with heavy weight design for continuous leaks and larger spills
  • Integrates safety signage and absorbency with bright high-visibility color
  • Allows for cost control and waste reduction through perforations
  • Warns of slips, trips, and falls with pre-printed message; bright color alerts to leaks even in low light