Learn More
MSC Brady SPC Drum Top Pads, Universal, Chemical, 22 in, 25/Pk
Catalog No. 5000802457
Encompass
These drum top pads are made from 100% meltblown polypropylene, suitable for universal and chemical applications. Designed to fit 55-gal drums, they protect floors by preventing pump splatters and drips. The pads are 22 in. in diameter and offer a total absorption capacity of 6 US gal per case.
  • Made from 100% meltblown polypropylene
  • Designed for universal and chemical applications
  • Offers a total package absorption capacity of 6 US gal
  • Measures 22 in. in diameter
  • Compatible with 55-gal drums featuring 1 and 2 holes
  • Protects floors by preventing pump splatters and drips
  • Complies with RoHS 2011/65/EU + (EU)2015/863 standards
Catalog No. 50-008-02457 Supplier MSC Supplier No. 00260513
These drum top pads are made from 100% meltblown polypropylene, suitable for universal and chemical applications. Designed to fit 55-gal drums, they protect floors by preventing pump splatters and drips. The pads are 22 in. in diameter and offer a total absorption capacity of 6 US gal per case.
  • Made from 100% meltblown polypropylene
  • Designed for universal and chemical applications
  • Offers a total package absorption capacity of 6 US gal
  • Measures 22 in. in diameter
  • Compatible with 55-gal drums featuring 1 and 2 holes
  • Protects floors by preventing pump splatters and drips
  • Complies with RoHS 2011/65/EU + (EU)2015/863 standards