Learn More
MSC Brady SPC MRO Plus Pad, Universal, 26 gal, 100/Case
Catalog No. 5000796213
Encompass
MRO Plus products are strong, durable, and low linting, making them ideal for almost any industrial setting. These highly versatile, gray absorbents are designed to pick up water-based, petroleum-based, and non-aggressive chemical fluids. The heavy-weight construction is specifically for continuous leaks and larger spills, which helps reduce material usage.
  • Absorbs up to 26 gal (US)
  • Perforated for cost control and waste reduction
  • Durable construction
  • Measures 15 in W x 19 in L
  • Heavy absorbency weight
  • Universal absorbency
  • Constructed with 3-ply meltblown polypropylene
  • Picks up water-based, petroleum-based, and non-aggressive chemical fluids
  • Low linting
  • Service temperature up to 160 °C (320 °F)
Catalog No. 50-007-96213 Supplier MSC Supplier No. 00260570
MRO Plus products are strong, durable, and low linting, making them ideal for almost any industrial setting. These highly versatile, gray absorbents are designed to pick up water-based, petroleum-based, and non-aggressive chemical fluids. The heavy-weight construction is specifically for continuous leaks and larger spills, which helps reduce material usage.
  • Absorbs up to 26 gal (US)
  • Perforated for cost control and waste reduction
  • Durable construction
  • Measures 15 in W x 19 in L
  • Heavy absorbency weight
  • Universal absorbency
  • Constructed with 3-ply meltblown polypropylene
  • Picks up water-based, petroleum-based, and non-aggressive chemical fluids
  • Low linting
  • Service temperature up to 160 °C (320 °F)