Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC Oil Absorbent Heavy Weight Pad, 1 side with blue coverstock

Catalog No. 17111125 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
24 gal./Bale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-125 24 gal./Bale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-125 Supplier Brady™ Supplier No. SPC105
Add to Cart
Add to Cart

SPC Oil-Absorbent Heavy Weight Pad absorbs petroleum-based liquids and repels water, making it ideal for outdoor marine or environmental applications, as well as for general industrial use.

  • White absorbents pick up petroleum-based fluids, paints and non-water soluble chemicals while repelling water - great for indoor and outdoor use
  • Heavy weight—specifically designed for continuous leaks and larger spills so you use less material
  • SPC™ absorbents are made with MAXX™ Technology for superior absorbency and greater strength without the added weight
  • Perforations provide cost control and reduce waste by allowing you to use only what you need

Specifications

Color White/Blue
Capacity (English) 24 gal./Bale
Material Polypropylene
Type SPC Oil Absorbent Heavyweight Pad, 1 side with blue coverstock
For Use With (Application) General industrial use, Marine and environmental spills, Oil refineries/Fuel chemical, General manufacturing
Packaging Quantity 100/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 38.1 x 48.2 cm (15 x 19 in.)
Length (Metric) 48.26 cm
Width (Metric) 38.1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.