Learn More
MSC Brady SPC Sorbent Roll, 15 in, 23 gal, 1 Each
Catalog No. 5000674517
Encompass
This sorbent roll is designed for universal use, capable of absorbing a variety of fluids including water, solvents, oil, fuel, coolants, and chemicals. Made from polypropylene, it measures 15 in wide and 150 ft long with a 23 gal capacity.
  • High durability
  • Two-ply construction
  • Top coverstock
  • Highly versatile
  • Durable
  • Camouflaged printed
  • Low linting
  • Absorbs water, solvents, oil, fuel, coolants, and chemicals
  • Heavy absorbency weight
  • Perforated
Catalog No. 50-006-74517 Supplier MSC Supplier No. 75535831
This sorbent roll is designed for universal use, capable of absorbing a variety of fluids including water, solvents, oil, fuel, coolants, and chemicals. Made from polypropylene, it measures 15 in wide and 150 ft long with a 23 gal capacity.
  • High durability
  • Two-ply construction
  • Top coverstock
  • Highly versatile
  • Durable
  • Camouflaged printed
  • Low linting
  • Absorbs water, solvents, oil, fuel, coolants, and chemicals
  • Heavy absorbency weight
  • Perforated