Learn More
MSC Brady SPC Sorbents Sock, Oil Only, 12 gal, 6/Pk.
Catalog No. 5000796339
Encompass
These polypropylene socks are specifically designed to absorb oil and oil-based fluids, suitable for both indoor and outdoor use. They are impervious to rot and mildew, and are unaffected by temperature. The socks feature strong wicking action, ensuring instant absorption upon contact with liquid without needing to be turned.
  • Designed to absorb oil and oil-based fluids
  • Suitable for indoor and outdoor use
  • Impervious to rot and mildew
  • Unaffected by temperature
  • Strong wicking action for instant absorption
Catalog No. 50-007-96339 Supplier MSC Supplier No. 09965740
These polypropylene socks are specifically designed to absorb oil and oil-based fluids, suitable for both indoor and outdoor use. They are impervious to rot and mildew, and are unaffected by temperature. The socks feature strong wicking action, ensuring instant absorption upon contact with liquid without needing to be turned.
  • Designed to absorb oil and oil-based fluids
  • Suitable for indoor and outdoor use
  • Impervious to rot and mildew
  • Unaffected by temperature
  • Strong wicking action for instant absorption