Learn More
MSC Brady SPC Sorbents Universal Spill Kit, 38 gal, 1 ea
Catalog No. 5000674519
Encompass
Brady SPC Sorbents universal spill kit is designed for quick spill response, capable of absorbing up to 38 gal. It is packaged in a 55 gal polyethylene drum and includes a variety of absorbents and safety gear.
  • Fifty 15 × 19 in pads
  • Four 3 in × 12 ft socks
  • Eight 18 × 18 in pillows
  • One pair goggles
  • One pair gloves
  • Five disposal bags
  • One emergency response guidebook
  • Universal kit type
  • Polyethylene drum container
  • Absorption capacity: 38 gal
  • Yellow color
Catalog No. 50-006-74519 Supplier MSC Supplier No. 75535856
Brady SPC Sorbents universal spill kit is designed for quick spill response, capable of absorbing up to 38 gal. It is packaged in a 55 gal polyethylene drum and includes a variety of absorbents and safety gear.
  • Fifty 15 × 19 in pads
  • Four 3 in × 12 ft socks
  • Eight 18 × 18 in pillows
  • One pair goggles
  • One pair gloves
  • Five disposal bags
  • One emergency response guidebook
  • Universal kit type
  • Polyethylene drum container
  • Absorption capacity: 38 gal
  • Yellow color