Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Static Resistant Heavy Pads

Catalog No. 17111140 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
16 gal./Bale
30 gal./Cs.
9 gal./Bale
Dimensions (L x W):
38.1 x 48.2 cm (15 x 19 in.)
76.2 x 76.2 cm (30 x 30 in.)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-140 9 gal./Bale 38.1 x 48.2 cm (15 x 19 in.)
17-111-119 30 gal./Cs. 38.1 x 48.2 cm (15 x 19 in.)
17-111-139 16 gal./Bale 76.2 x 76.2 cm (30 x 30 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-111-140 Supplier Brady™ Supplier No. SR1850
Only null left
Add to Cart
Add to Cart

For use with oil and petroleum-based fluids

  • Treated to resist static electricity build-up
  • Absorb only oil and not water
  • Single-ply
  • Color: White
  • Meets National Fire Protection Association (NFPA) specifications

Specifications

Color White
Capacity (English) 9 gal./Bale
Material Polypropylene
Type Static-resistant Heavy Pad
For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
Packaging Quantity 50/Cs.
Dimensions (L x W) 38.1 x 48.2 cm (15 x 19 in.)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.