Learn More
Brady™ Spill control Putty Patty
Catalog No. 17111086 Shop All Brady Corporation Products
Change view
Click to view available options
Type:
Spill Control Putty Patty
Catalog No. Type
17-111-086 Spill Control Putty Patty
1 options
Catalog No. 17-111-086 Supplier Brady™ Supplier No. PUTPAT
Add to Cart
Add to Cart
Spill Control Putty Patty is designed for immediate control of fuels and hazardous material leaks.
  • Sticks to dirty, rusty or greasy surfaces
  • Apply directly over and around the leak; work into the hole and press down for good adhesion
  • No surface preparation necessary
Specifications
Type Spill Control Putty Patty
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov