Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Spill Deck

Catalog No. 17111229 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17-111-229 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-229 Supplier Brady™ Supplier No. SCSD2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Spill Deck contains potentially hazardous leaks in drum storage and dispensing areas.

  • Injection molding ensures exceptional durability and consistent specifications
  • Manufactured to standard lip and grate heights to allow uninhibited movement
  • Low profile, 21-gallon capacity, two-drum spill deck contains hazardous leaks in drum storage and dispensing areas
  • Two-drum deck includes removable grate and built-in clips allowing for easy connection and expansion - additional decks and accessories sold separately

Specifications

Color Blue/Yellow
Material Polyethylene
Type Drum Spill Deck
Dimensions (L x W x H) 132 x 66 x 15.2 cm (52 x 26 x 6 in.)
Length (Metric) 132.08 cm
Width (Metric) 66.04 cm
Weight (English) 30.8 lb.
Weight (Metric) 67.91 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.