Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Spill Kaddie Refill, Universal

Catalog No. 17111250 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
22 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-250 22 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-250 Supplier Brady™ Supplier No. SKAK2R
Add to Cart
Add to Cart

Universal Spill Kaddie Refill is intended for use with the Allwik™ universal spill kaddie. (Refill kit only—container sold separately.)

  • Universal absorbency - picks up water-based, petroleum-based and non-aggressive, chemical fluids
  • Absorbs up to 22 gallons per kit
  • RoHS Compatibility Pass 2011/65/EU

Specifications

Color Gray
Capacity (English) 22 gal.
Capacity (Metric) 83.2 L
Type Spill Kaddie Refill, Universal
For Use With (Application) Universal - All purpose spills
Includes (1) 15 in. x 73 ft. Roll, (5) 3 in. x 4 ft. socks, (8) 10 x 10 in. pillows, 1 pair Nitrile gloves, 1 pair goggles, 5 disposal bags, handbook
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.