Learn More
Brady™ Spill Response Plus Oil Only Absorbents
Catalog No. 19140737 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
19-140-737 Polypropylene
1 options
Catalog No. 19-140-737 Supplier Brady™ Supplier No. SRPO200
  • $130.00
    $90.65 / Case of 200
    Save $39.35
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Absorb just about any oil based liquid
  • For preventing slips, trips and falls that commonly occur in the retail, hospitality and grocery markets
  • Dimpled design to be highly absorbent, durable and reduce waste
  • Absorbs paints, vegetable oils and other non-water soluble oil-based fluids
  • Convenient size makes it ideal for employees to fold and carry in their pockets for quick response to spills
  • Available as pads
  • Specifications
    Material Polypropylene
    Type Absorbent Pad, Oil-only
    Packaging Quantity 200/Cs.