Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Striped Warning Tapes

Catalog No. 19103762 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, White
Black, Yellow
Green, White
Red, White
Width (English):
1 in.
1.5 in.
2 in.
3 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Width (English)
19-103-762 Black, Yellow 1 in.
19-103-763 Black, Yellow 1.5 in.
19-103-815 Green, White 2 in.
19-103-758 Black, White 3 in.
19-103-760 Red, White 2 in.
19-103-761 Red, White 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-103-762 Supplier Brady™ Supplier No. 55300
Add to Cart
Add to Cart

Indicates hazardous areas clearly, marks physical hazards

  • Permanent adhesive stands up to tough indoor conditions
  • Colors are sealed beneath protective layer of abrasion resistant clear vinyl
  • Holds on to almost any clean, dry surface

Specifications

Adhesive Type Rubber-based Pressure-Sensitive
Color Black, Yellow
Length (English) 54 ft.
Length (Metric) 16.4 m
Width (English) 1 in.
Width (Metric) 2.54 cm
Material B-950 Vinyl
Type Striped Warning Tape
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.