Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Striper Paint

Catalog No. 19807090 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-807-090 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-807-090 Supplier Brady™ Supplier No. 56901

missing translation for 'validMainframeMsg'

Aisle Marking Paint is a quick, easy way to identify aisles and passageways in your facility.

  • Colorful Aisle Marking Paint is used with paint striper sold previous catalogs (paint does not work with Universal Line Applicator). Standard package: case of twelve 20 fl. oz. cans.

Specifications

Capacity (English) 20 oz.
Product Type Striper Paint
Color Yellow
Capacity (Metric) 591.4 mL
For Use With (Equipment) For use with Colorful Aisle Marking Paint
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.