Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ToughStripe™ 'Do Not Enter'Floor Sign

Catalog No. 19150825 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-150-825 Black on White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-825 Supplier Brady™ Supplier No. 104497
  • $110.00
    $67.65 / Each of 1

    Save $42.35

    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Access and admittance signs identify restricted areas or alert employees of potential hazards. These signs also communicate directions, safety warning and private property notices.

  • ToughStripe floor signs feature durable floor marking tape with a high-performance adhesive
  • Sign reads “DO NOT ENTER” (with symbols and anti-skid areas)
  • Red and black on white
  • Size: 35.56H × 45.72Wcm (14H × 18W in.)
  • B-534 polyester — surface printed polyester with clear polyester overlaminate, anti-skid areas and rubber-based adhesive

  • Bright colors and bold text provide high visibility to critical messages
  • Designed to withstand harsh industrial environments

Specifications

Certifications/Compliance ASTM ANSI F1679, PSTC-33
Material Polyester
Quantity 1 Ea.
Mounting Type Self-adhesive
For Use With (Application) Used to mark aisles, passageways, storage locations, point out safety equipment and exits and enforce protective equipment requirements
Height (English) 14 in.
Max. Temperature (English) 130°F
Min. Temperature (English) 0°F
Thickness 0.01 in.
Width (English) 18 in.
Temperature Range (Metric) -18°C to 54°C
Shape Rectangular
Temperature Range (English) 0°F to 130°F
Languages English
Displayed Text Do Not Enter
Product Type Floor Sign
Color Black on White
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.