Learn More
MSC Brady ToughStripe Floor Marking Dots, Red Polyester, 3.5 in, 350/Roll
Catalog No. 5000796740
Encompass
ToughStripe die-cut floor marking dots are constructed from rugged polyester with an ultra-aggressive adhesive, designed to withstand heavy forklift traffic and enhance industrial safety and organization.
  • High-gloss surface maintains a new paint-like appearance
  • Withstands forklift traffic without tearing or lifting
  • Ideal for marking aisles, passageways, and storage locations
  • Can be easily applied by one person
Catalog No. 50-007-96740 Supplier MSC Supplier No. 12282034
ToughStripe die-cut floor marking dots are constructed from rugged polyester with an ultra-aggressive adhesive, designed to withstand heavy forklift traffic and enhance industrial safety and organization.
  • High-gloss surface maintains a new paint-like appearance
  • Withstands forklift traffic without tearing or lifting
  • Ideal for marking aisles, passageways, and storage locations
  • Can be easily applied by one person