Learn More
Brady™ ToughStripe™ Striped Tape
Catalog No. 19103740 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Green, White
Catalog No. Color
19-103-740 Green, White
1 options
Catalog No. 19-103-740 Supplier Brady™ Supplier No. 132436
  • $105.50
    $63.65 / Each of 1
    Save $41.85
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For marking aisles, passageways and storage locations
  • Points out important safety equipment and egress paths
  • Exceeds safety standards for clean, dry non-slip surfaces
  • 2mm thickness
  • Resists harsh chemicals
Specifications
Adhesive Type Permanent Rubber-based
Dimensions (L x W) 100 ft. x 2 in.
Color Green, White
Certifications/Compliance ANSI A1264.2-2006 and OSHA 1910.22
Material Polyester
Type Striped Tape