Learn More
Brady™ TrackMat™ Heavy Pad Absorbent
Catalog No. 17111098 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
18 gal.
Catalog No. Capacity (English)
17-111-098 18 gal.
1 options
Catalog No. 17-111-098 Supplier Brady™ Supplier No. TM30
For high volume petroleum spills
  • Three-ply construction
  • UV-resistant top layer
  • Features polypropylene core
  • Non-leaking and non-slip barrier packing
Specifications
Capacity (English) 18 gal.
Type TrackMat Heavy Pad Absorbent
Dimensions (L x W) 73.6 x 73.6 cm (23 x 23 in.)