Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Universal All-Purpose Spill Control Absorbent Roll

Catalog No. 17111007 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
27 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-007 27 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-007 Supplier Brady™ Supplier No. UN75200

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

All-purpose sorbent made to handle virtually any liquid (water, petroleum and chemical-based)

Specifications

Capacity (English) 27 gal.
Capacity (Metric) 102.2 L
Type Universal Spill Control Absorbent Roll
Packaging Quantity 2 Rolls
Dimensions (L x W) Roll: 200 ft. x 7.5 in.
Width (English) 7.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.