Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Universal Plus Pads

Catalog No. 19150258 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
12 x 12 in.
19 x 17 in.
Packaging Quantity:
100/Pk.
200/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Packaging Quantity
19-150-258 19 x 17 in. 100/Pk.
19-150-257 12 x 12 in. 200/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-150-258 Supplier Brady™ Supplier No. SPCUN1719
  • $790.00
    $635.65 / Case of 100

    Save $154.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color-coded chemical absorbents for safe clean-up of hazardous fluids

  • Chemically inert and ideal for aggressive fluids
  • Intended for use with chemicals and hazardous materials
  • Universal meltblown polypropylene
  • Three-ply construction makes it stronger, more durable and reduces lint
  • Dimpled
  • Medium weight

SPCUN1719 is perforated.

†Perforated

Specifications

Certifications/Compliance RoHS
Color Green
Capacity (English) 20.5 gal.
Capacity (Metric) 77.6 L
Material Polypropylene
Type Universal Plus Pad
For Use With (Application) Aggressive chemicals and hazardous fluids
Packaging Quantity 100/Pk.
Thickness 3-ply
Weight Capacity Medium-weight
Dimensions (L x W) 19 x 17 in.
Length (English) 19 in.
Length (Metric) 48.2 cm
Width (English) 17 in.
Width (Metric) 43 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.