Learn More
Brady™ UXT Xtra Tough Universal Heavy Rolls
Catalog No. 17111195 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
17 gal./Bale
Catalog No. Capacity (English)
17-111-195 17 gal./Bale
1 options
Catalog No. 17-111-195 Supplier Brady™ Supplier No. UXT15P
  • $167.00
    $118.65 / Each of 1
    Save $48.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Made to handle virtually any liquid (water, petroleum and chemical-based)
  • Three-ply construction
  • Double-sided, low-linting coverstock and meltblown core
  • Abrasion resistant and non-abrasive performance
  • Perforated
  • Specifications
    Capacity (English) 17 gal./Bale
    Material Polypropylene
    Type UXT Xtra Tough Universal Heavy Roll
    For Use With (Application) Food Processing, High Tech Industries, Machining of small parts, Paint room, Pharmaceutical, Printing
    Packaging Quantity 1 Ea.
    Weight Capacity Heavyweight
    Dimensions (L x W) 45.72 m x 38.1 cm (150 ft. x 15 in.)
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov